Learn More
Medchemexpress LLC HY-18619 5mg Medchemexpress, YL-109 CAS:36341-25-0 Purity:>98%
SDP
Catalog No. 502082057
Encompass_Preferred
Medchemexpress, HY-18619 5mg YL-109 CAS:36341-25-0 YL-109 is an antitumor agent that can induce carboxyl terminus of Hsp70-interacting protein (CHIP) expression through aryl hydrocarbon receptor (AhR) signaling. YL-109 has ability to inhibit breast cancer cell growth and invasiveness. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2057 Supplier Medchemexpress LLC Supplier No. HY186195MG
Medchemexpress, HY-18619 5mg YL-109 CAS:36341-25-0 YL-109 is an antitumor agent that can induce carboxyl terminus of Hsp70-interacting protein (CHIP) expression through aryl hydrocarbon receptor (AhR) signaling. YL-109 has ability to inhibit breast cancer cell growth and invasiveness. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.