Learn More
Medchemexpress LLC YL-5092 | 3056857-07-6 | 99.8% | 441.43 | 1 ML
SDP
Catalog No. 5000391732
Encompass_Preferred
YL-5092 is a selective YT521-B homology (YTH) domain-containing protein 1 (YTHDC1) inhibitor with an IC50 of 7.4 nM and a KD of 29.6 nM. It can suppress cancer cell proliferation and induce cell G0/G1 phase arrest and apoptosis. YL-5092 is used for cancer research, including acute myeloid leukemia (AML).
  • Selectively inhibits YT521-B homology (YTH) domain-containing protein 1.
  • Suppresses cancer cell proliferation.
  • Induces cell G0/G1 phase arrest.
  • Promotes apoptosis in cancer cells.
  • Impairs leukemogenesis and improves survival rates in mouse tumor models.
  • Reduces leukemia infiltration in peripheral blood, spleen, and bone marrow.
  • Decreases the expression of YTHDC1 target genes like MYC, FOXM1, GINS1, MCM2, MCM4, and MCM5.
Catalog No. 50-003-91732 Supplier Medchemexpress LLC Supplier No. HY16460710MM/1ML
YL-5092 is a selective YT521-B homology (YTH) domain-containing protein 1 (YTHDC1) inhibitor with an IC50 of 7.4 nM and a KD of 29.6 nM. It can suppress cancer cell proliferation and induce cell G0/G1 phase arrest and apoptosis. YL-5092 is used for cancer research, including acute myeloid leukemia (AML).
  • Selectively inhibits YT521-B homology (YTH) domain-containing protein 1.
  • Suppresses cancer cell proliferation.
  • Induces cell G0/G1 phase arrest.
  • Promotes apoptosis in cancer cells.
  • Impairs leukemogenesis and improves survival rates in mouse tumor models.
  • Reduces leukemia infiltration in peripheral blood, spleen, and bone marrow.
  • Decreases the expression of YTHDC1 target genes like MYC, FOXM1, GINS1, MCM2, MCM4, and MCM5.