Learn More
Medchemexpress LLC Yl-5092 | 3056857-07-6 | 99.83% | 441.43 | 50 MG
SDP
Catalog No. 5000391635
Encompass_Preferred
YL-5092 is a selective YT521-B homology (YTH) domain-containing protein 1 (YTHDC1) inhibitor with an IC50 of 7.4 nM and a KD of 29.6 nM. It can suppress cancer cell proliferation and induce cell G0/G1 phase arrest and apoptosis. YL-5092 can be used for the research of cancer, such as acute myeloid leukemia (AML).
  • Selective YTHDC1 inhibitor with IC50 of 7.4 nM and KD of 29.6 nM.
  • Suppresses cancer cell proliferation.
  • Induces cell G0/G1 phase arrest and apoptosis.
  • Applicable for cancer research, including acute myeloid leukemia (AML).
Catalog No. 50-003-91635 Supplier Medchemexpress LLC Supplier No. HY16460750MG
Add to Cart
Add to Cart
YL-5092 is a selective YT521-B homology (YTH) domain-containing protein 1 (YTHDC1) inhibitor with an IC50 of 7.4 nM and a KD of 29.6 nM. It can suppress cancer cell proliferation and induce cell G0/G1 phase arrest and apoptosis. YL-5092 can be used for the research of cancer, such as acute myeloid leukemia (AML).
  • Selective YTHDC1 inhibitor with IC50 of 7.4 nM and KD of 29.6 nM.
  • Suppresses cancer cell proliferation.
  • Induces cell G0/G1 phase arrest and apoptosis.
  • Applicable for cancer research, including acute myeloid leukemia (AML).