Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC YL-939 | 3023925-68-7 | 99.0% | 426.51 g/mol | C25H26N6O | 10 MG
SDP

Catalog No. 5000287515
Encompass_Preferred

YL-939 is a small-molecule ferroptosis inhibitor that prevents iron-dependent cell death by modulating the PHB2/ferritin/iron axis. It has been characterized as a non-classical ferroptosis inhibitor with demonstrated protective activity in cellular assays.

  • Inhibits ferroptosis via the PHB2/ferritin/iron axis.
  • Demonstrated cellular activity in vitro against ferroptotic insults.
  • High chemical purity (~99.0%) and defined physical properties.
  • Off-white to light-yellow solid, molecular weight 426.51 g/mol.
  • CAS number 3023925-68-7 for unambiguous identification.
  • Soluble in DMSO (100 mg/mL) and amenable to in vivo formulations.
  • Store as powder at -20°C; limited stability in solution.

Catalog No. 50-002-87515 Supplier Medchemexpress LLC Supplier No. HY15209310MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

YL-939 is a small-molecule ferroptosis inhibitor that prevents iron-dependent cell death by modulating the PHB2/ferritin/iron axis. It has been characterized as a non-classical ferroptosis inhibitor with demonstrated protective activity in cellular assays.

  • Inhibits ferroptosis via the PHB2/ferritin/iron axis.
  • Demonstrated cellular activity in vitro against ferroptotic insults.
  • High chemical purity (~99.0%) and defined physical properties.
  • Off-white to light-yellow solid, molecular weight 426.51 g/mol.
  • CAS number 3023925-68-7 for unambiguous identification.
  • Soluble in DMSO (100 mg/mL) and amenable to in vivo formulations.
  • Store as powder at -20°C; limited stability in solution.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.