Learn More
Medchemexpress LLC YL-939 | 3023925-68-7 | C25H26N6O | 100 MG
SDP
Catalog No. 5000373731
Encompass_Preferred
YL-939 is a potent ferroptosis inhibitor. It inhibits ferroptosis by targeting the PHB2/ferritin/iron axis.
  • Efficiently protects cells from ferroptosis with varying IC50 values across different cell lines.
  • Reduces the level of reactive oxygen species in cytosolic and membrane lipids.
  • Improves ferritin protein expression and blocks autophagosomes/lysosomes, inhibiting ferritinophagy.
  • Ameliorates liver damage in an acetaminophen-induced acute liver injury model.
  • Inhibits cell death and inflammatory infiltration in liver tissues.
Catalog No. 50-003-73731 Supplier Medchemexpress LLC Supplier No. HY152093100MG
YL-939 is a potent ferroptosis inhibitor. It inhibits ferroptosis by targeting the PHB2/ferritin/iron axis.
  • Efficiently protects cells from ferroptosis with varying IC50 values across different cell lines.
  • Reduces the level of reactive oxygen species in cytosolic and membrane lipids.
  • Improves ferritin protein expression and blocks autophagosomes/lysosomes, inhibiting ferritinophagy.
  • Ameliorates liver damage in an acetaminophen-induced acute liver injury model.
  • Inhibits cell death and inflammatory infiltration in liver tissues.