Learn More
Medchemexpress LLC YL-939 | 3023925-68-7 | C25H26N6O | 5 MG
SDP
Catalog No. 5000374134
Encompass_Preferred
YL-939 is a potent ferroptosis inhibitor. YL-939 inhibits ferroptosis by targeting the PHB2/ferritin/iron axis.
  • Potent ferroptosis inhibitor
  • Inhibits ferroptosis by targeting the PHB2/ferritin/iron axis
Catalog No. 50-003-74134 Supplier Medchemexpress LLC Supplier No. HY1520935MG
YL-939 is a potent ferroptosis inhibitor. YL-939 inhibits ferroptosis by targeting the PHB2/ferritin/iron axis.
  • Potent ferroptosis inhibitor
  • Inhibits ferroptosis by targeting the PHB2/ferritin/iron axis