Learn More
Medchemexpress LLC HY-15840 5mg Medchemexpress, YLF-466D CAS:1273323-67-3 Purity:>98%
SDP
Catalog No. 502081803
Encompass_Preferred
Medchemexpress, HY-15840 5mg YLF-466D CAS:1273323-67-3 YLF-466D is a newly developed AMPK activator, which inhibits platelet aggregation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1803 Supplier Medchemexpress LLC Supplier No. HY158405MG
Medchemexpress, HY-15840 5mg YLF-466D CAS:1273323-67-3 YLF-466D is a newly developed AMPK activator, which inhibits platelet aggregation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.