Learn More
Andwin Scientific YLW TPS/53508-810 10000/CS
SDP
Catalog No. NC0400458
Encompass_Preferred
The Signature™ Bevel Point Pipet Tips offer maximum sample recovery and minimal fluid retention. This product features compatibility most pipettors. The tips are autoclavable up to 122C (252F) and its beveled ends provide better accuracy and reprodu
Catalog No. NC0400458 Supplier Andwin Scientific Supplier No. 53508810CS
The Signature™ Bevel Point Pipet Tips offer maximum sample recovery and minimal fluid retention. This product features compatibility most pipettors. The tips are autoclavable up to 122C (252F) and its beveled ends provide better accuracy and reprodu