Learn More
Abcam YM-155, survivin inhibitor, 5MG
Catalog No. 502702899
Encompass_Preferred
MW 443.3 Da, Purity >98%. Novel small molecule potent survivin suppressant. Inhibits survivin promoter activity (IC₅₀ = 0.54 nM) and down-regulates survivin in vitro and in vivo. Induces apoptosis in human tumor cell lines in vitro at 10 nM, with little effect on other IAP or Bcl-2 family members.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-2899 Supplier Abcam Supplier No. AB2223765MG
MW 443.3 Da, Purity >98%. Novel small molecule potent survivin suppressant. Inhibits survivin promoter activity (IC₅₀ = 0.54 nM) and down-regulates survivin in vitro and in vivo. Induces apoptosis in human tumor cell lines in vitro at 10 nM, with little effect on other IAP or Bcl-2 family members.
The product is subject to the following: Abcam Restricted Use Statement