Learn More
Cayman Chemical YM-201636, 371942-69-7, 5 mg
SDP
Supplier:  Cayman Chemical 135765
Encompass_Preferred
A cell-permeable and selective inhibitor of PIKfyve (IC50 = 33 nM); reversibly impairs endosomal trafficking in NIH3T3 cells and blocks retroviral exit by budding from cells; inhibits basal and insulin-activated 2-deoxyglucose uptake (IC50 = 54 nM) in adipocytes
Catalog No. NC1313687