Learn More
Medchemexpress LLC HY-13228 5mg Medchemexpress, YM-201636 CAS:371942-69-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY132285MG
Encompass_Preferred
Medchemexpress, HY-13228 5mg YM-201636 CAS:371942-69-7 YM-201636 is a potent and selective PIKfyve inhibitor with an IC50 of 33 nM. YM-201636 also inhibits p110α with an IC50 of 3.3 μM. YM-201636 inhibits retroviral replication. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0264
 Restricted Item