Learn More
Medchemexpress LLC YM-244769 | 838819-70-8 | 99.2% | 443.47 | 50 MG
SDP
Catalog No. 5000348181
Encompass_Preferred
YM-244769 is a potent, selective, and orally active Na+/Ca2+ exchanger (NCX) inhibitor. It preferentially inhibits NCX3 and suppresses the unidirectional outward NCX current (Ca2+ entry mode) with IC50s of 18 nM and 50 nM, respectively. It efficiently protects against hypoxia/reoxygenation-induced SH-SY5Y neuronal cell damage and can increase urine volume and urinary excretion of electrolytes in mice.
  • Potent, selective, and orally active Na+/Ca2+ exchanger (NCX) inhibitor.
  • Preferentially inhibits NCX3 (IC50 = 18 nM).
  • Suppresses unidirectional outward NCX current (Ca2+ entry mode) with an IC50 of 50 nM.
  • Efficiently protects against hypoxia/reoxygenation-induced SH-SY5Y neuronal cell damage.
  • Increases urine volume and urinary excretion of electrolytes in mice.
Catalog No. 50-003-48181 Supplier Medchemexpress LLC Supplier No. HY136182A50MG
YM-244769 is a potent, selective, and orally active Na+/Ca2+ exchanger (NCX) inhibitor. It preferentially inhibits NCX3 and suppresses the unidirectional outward NCX current (Ca2+ entry mode) with IC50s of 18 nM and 50 nM, respectively. It efficiently protects against hypoxia/reoxygenation-induced SH-SY5Y neuronal cell damage and can increase urine volume and urinary excretion of electrolytes in mice.
  • Potent, selective, and orally active Na+/Ca2+ exchanger (NCX) inhibitor.
  • Preferentially inhibits NCX3 (IC50 = 18 nM).
  • Suppresses unidirectional outward NCX current (Ca2+ entry mode) with an IC50 of 50 nM.
  • Efficiently protects against hypoxia/reoxygenation-induced SH-SY5Y neuronal cell damage.
  • Increases urine volume and urinary excretion of electrolytes in mice.