Learn More
Medchemexpress LLC Ym-244769 dihydrochloride | 1780390-65-9 | 516.39 | 25 MG
SDP
Catalog No. 5000351858
Encompass_Preferred
Ym-244769 dihydrochloride is a potent, selective, and orally active Na+/Ca2+ exchanger (NCX) inhibitor. It preferentially inhibits NCX3 and suppresses the unidirectional outward NCX current (Ca2+ entry mode), with IC50 values of 18 nM and 50 nM, respectively. This compound efficiently protects against hypoxia/reoxygenation-induced SH-SY5Y neuronal cell damage. It can also increase urine volume and urinary excretion of electrolytes in mice.
  • Potent and selective Na+/Ca2+ exchanger (NCX) inhibitor.
  • Preferentially inhibits NCX3.
  • Suppresses unidirectional outward NCX current (Ca2+ entry mode).
  • Protects against hypoxia/reoxygenation-induced neuronal cell damage.
  • Increases urine volume and urinary excretion of electrolytes.
Catalog No. 50-003-51858 Supplier Medchemexpress LLC Supplier No. 25MGHY136182
Ym-244769 dihydrochloride is a potent, selective, and orally active Na+/Ca2+ exchanger (NCX) inhibitor. It preferentially inhibits NCX3 and suppresses the unidirectional outward NCX current (Ca2+ entry mode), with IC50 values of 18 nM and 50 nM, respectively. This compound efficiently protects against hypoxia/reoxygenation-induced SH-SY5Y neuronal cell damage. It can also increase urine volume and urinary excretion of electrolytes in mice.
  • Potent and selective Na+/Ca2+ exchanger (NCX) inhibitor.
  • Preferentially inhibits NCX3.
  • Suppresses unidirectional outward NCX current (Ca2+ entry mode).
  • Protects against hypoxia/reoxygenation-induced neuronal cell damage.
  • Increases urine volume and urinary excretion of electrolytes.