Learn More
Medchemexpress LLC Ym-244769 dihydrochloride | 1780390-65-9 | MFCD28160863 | ≥98.0% | C26H22FN3O3·2HCl | 10MG
SDP
Catalog No. 5000260216
Encompass_Preferred
YM-244769 dihydrochloride is a research reagent that potently and selectively inhibits the Na+/Ca2+ exchanger (NCX). Supplied as the dihydrochloride salt, it is intended for cellular and biochemical studies probing NCX function.
  • Potent, selective Na+/Ca2+ exchanger (NCX) inhibitor.
  • Supplied as the dihydrochloride salt for improved solubility.
  • Suitable for cellular and biochemical assays.
  • High purity (≥98.0% by HPLC).
  • Characterized with CAS 1780390-65-9 for unambiguous identification.
Catalog No. 50-002-60216 Supplier Medchemexpress LLC Supplier No. HY13618210MG
YM-244769 dihydrochloride is a research reagent that potently and selectively inhibits the Na+/Ca2+ exchanger (NCX). Supplied as the dihydrochloride salt, it is intended for cellular and biochemical studies probing NCX function.
  • Potent, selective Na+/Ca2+ exchanger (NCX) inhibitor.
  • Supplied as the dihydrochloride salt for improved solubility.
  • Suitable for cellular and biochemical assays.
  • High purity (≥98.0% by HPLC).
  • Characterized with CAS 1780390-65-9 for unambiguous identification.