Learn More
Medchemexpress LLC YM-244769 hydrochlor 10mg | 837424-39-2 | 10MG
SDP
Catalog No. 5000240308
Encompass_Preferred
YM-244769 hydrochloride is a potent selective and orally active Na /Ca2 exchanger (NCX) inhibitor YM-244769 hydrochloride preferentially inhibits NCX3 and suppresses the unidirectional outward NCX current (Ca2 entry mode) with IC50s of 18 nM and 50 nM respectively YM-244769 hydrochloride efficiently protects against hypoxia/reoxygenation-induced SH-SY5Y neuronal cell damage YM-244769 hydrochloride can also increase urine volume and urinary excretion of electrolytes in mice[1 [2 [3
Catalog No. 50-002-40308 Supplier Medchemexpress LLC Supplier No. HY10765910MG
YM-244769 hydrochloride is a potent selective and orally active Na /Ca2 exchanger (NCX) inhibitor YM-244769 hydrochloride preferentially inhibits NCX3 and suppresses the unidirectional outward NCX current (Ca2 entry mode) with IC50s of 18 nM and 50 nM respectively YM-244769 hydrochloride efficiently protects against hypoxia/reoxygenation-induced SH-SY5Y neuronal cell damage YM-244769 hydrochloride can also increase urine volume and urinary excretion of electrolytes in mice[1 [2 [3