Learn More
Medchemexpress LLC YM-244769 hydrochloride | 837424-39-2 | 99.0% | 479.93 g·mol⁻1 | C26H23ClFN3O3 | 5 MG
SDP
Catalog No. 5000240250
Encompass_Preferred
YM-244769 hydrochloride is a potent, selective, and orally active Na+/Ca2+ exchanger (NCX) inhibitor used in preclinical research. It preferentially inhibits NCX3 with low-nanomolar activity and has demonstrated neuroprotective effects in cell models and effects on urine volume and electrolyte excretion in animal studies.
  • Potent, selective inhibition of Na+/Ca2+ exchanger (NCX3) with low-nanomolar potency.
  • Orally active pharmacology suitable for in vivo preclinical studies.
  • Demonstrated neuroprotective activity in hypoxia/reoxygenation cell models.
  • Shown to increase urine volume and electrolyte excretion in animal studies.
  • Provided as a high-purity solid with recommended cold storage for stability.
Catalog No. 50-002-40250 Supplier Medchemexpress LLC Supplier No. HY1076595MG
YM-244769 hydrochloride is a potent, selective, and orally active Na+/Ca2+ exchanger (NCX) inhibitor used in preclinical research. It preferentially inhibits NCX3 with low-nanomolar activity and has demonstrated neuroprotective effects in cell models and effects on urine volume and electrolyte excretion in animal studies.
  • Potent, selective inhibition of Na+/Ca2+ exchanger (NCX3) with low-nanomolar potency.
  • Orally active pharmacology suitable for in vivo preclinical studies.
  • Demonstrated neuroprotective activity in hypoxia/reoxygenation cell models.
  • Shown to increase urine volume and electrolyte excretion in animal studies.
  • Provided as a high-purity solid with recommended cold storage for stability.