Learn More
Medchemexpress LLC Ym-244769 (hydrochloride) | 837424-39-2 | MFCD31561984 | 99.0% | 479.93 g/mol | C26H23ClFN3O3 | 50 MG
SDP
Catalog No. 5000240261
Encompass_Preferred
YM-244769 hydrochloride is a potent, selective inhibitor of the Na+/Ca2+ exchanger (NCX) supplied as a high-purity research chemical for pharmacological and physiological studies. The product is provided as a hydrochloride salt in powder form and is used in cardiovascular and neurological research to probe NCX-mediated transport and signaling.
  • Potent, selective inhibition of Na+/Ca2+ exchanger for NCX studies.
  • High purity suitable for in vitro and in vivo research.
  • Provided as a stable hydrochloride salt for reproducible dosing.
  • Supplied as a powder for flexible formulation and storage.
  • Documented certificate of analysis and safety data sheet available for quality assurance.
Catalog No. 50-002-40261 Supplier Medchemexpress LLC Supplier No. HY10765950MG
YM-244769 hydrochloride is a potent, selective inhibitor of the Na+/Ca2+ exchanger (NCX) supplied as a high-purity research chemical for pharmacological and physiological studies. The product is provided as a hydrochloride salt in powder form and is used in cardiovascular and neurological research to probe NCX-mediated transport and signaling.
  • Potent, selective inhibition of Na+/Ca2+ exchanger for NCX studies.
  • High purity suitable for in vitro and in vivo research.
  • Provided as a stable hydrochloride salt for reproducible dosing.
  • Supplied as a powder for flexible formulation and storage.
  • Documented certificate of analysis and safety data sheet available for quality assurance.