Learn More
AdipoGen YM-254890, 568580-02-9, 500mg
SDP
Supplier:  AdipoGen AGCN20509MC05
Encompass_Preferred
YM-254890 (500 g), AdipoGen Life Sciences. Chemical. CAS: 568580-02-9. Formula: C46H69N7O15. MW: 960.1. Cyclic depsipeptide composed of unique amino acids differing from normal amino acids. Membrane permeable, potent and selective Galphaq family inhibitor. Inhibits the signal transduction of Galphaq, Galpha11 and Galpha14 (IC50=0.095M) by blocking the exchange of GDP for GTP, preventing the activation of the G protein. Galphaq signaling has been shown to regulate brown/beige adipocytes using the structurally similar specific Galphaq family inhibitor FR900359. It is suggested that inhibition of this pathway may be a novel therapeutic approach to combat obesity.
Catalog No. 50-149-0016