Learn More
Medchemexpress LLC YM-298198 hydrochloride | 1216398-09-2 | MFCD08703122 | 98.0% | 378.92 g/mol | C18H23ClN4OS | 50 MG
SDP
Catalog No. 5000244996
Encompass_Preferred
YM-298198 hydrochloride is the hydrochloride salt of YM-298198, a selective, orally active, non-competitive antagonist of metabotropic glutamate receptor 1 (mGluR1). Supplied as a solid research-grade reagent, it is used in pharmacology studies and neurological disease models.
  • Selective mGluR1 antagonist for receptor pharmacology studies.
  • Orally active and non-competitive mechanism of action.
  • Supplied as a solid hydrochloride salt for stability and handling.
  • Molecular formula C18H23ClN4OS and molecular weight 378.92 g/mol.
  • High purity suitable for research applications.
Catalog No. 50-002-44996 Supplier Medchemexpress LLC Supplier No. HY10356850MG
YM-298198 hydrochloride is the hydrochloride salt of YM-298198, a selective, orally active, non-competitive antagonist of metabotropic glutamate receptor 1 (mGluR1). Supplied as a solid research-grade reagent, it is used in pharmacology studies and neurological disease models.
  • Selective mGluR1 antagonist for receptor pharmacology studies.
  • Orally active and non-competitive mechanism of action.
  • Supplied as a solid hydrochloride salt for stability and handling.
  • Molecular formula C18H23ClN4OS and molecular weight 378.92 g/mol.
  • High purity suitable for research applications.