Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC YM-298198 hydrochloride | 1216398-09-2 | MFCD08703122 | ≥99.0% | 378.92 | C18H23ClN4OS | 10 MG
SDP

Catalog No. 5000245160
Encompass_Preferred

YM-298198 hydrochloride is the hydrochloride salt of a selective, high-affinity, non-competitive antagonist of metabotropic glutamate receptor 1 (mGluR1) supplied for research use in small quantities.

  • Selective mGluR1 antagonist with Ki = 19 nM and IC50 = 16 nM in functional assays.
  • High reported purity (≥99.0% for latest batches).
  • Molecular formula C18H23ClN4OS and molecular weight 378.92.
  • CAS number 1216398-09-2 for substance identification.
  • Available in small pack sizes such as 10 mg for research applications.
  • Recommended storage: -20°C sealed; in solvent: -80°C up to 6 months or -20°C up to 1 month, protected from moisture.

Catalog No. 50-002-45160 Supplier Medchemexpress LLC Supplier No. HY10356810MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

YM-298198 hydrochloride is the hydrochloride salt of a selective, high-affinity, non-competitive antagonist of metabotropic glutamate receptor 1 (mGluR1) supplied for research use in small quantities.

  • Selective mGluR1 antagonist with Ki = 19 nM and IC50 = 16 nM in functional assays.
  • High reported purity (≥99.0% for latest batches).
  • Molecular formula C18H23ClN4OS and molecular weight 378.92.
  • CAS number 1216398-09-2 for substance identification.
  • Available in small pack sizes such as 10 mg for research applications.
  • Recommended storage: -20°C sealed; in solvent: -80°C up to 6 months or -20°C up to 1 month, protected from moisture.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.