Learn More
Medchemexpress LLC YM-341619 | 643082-52-4 | 98.0% | C22H21F3N6O2 | 10MG
SDP
Catalog No. 5000261283
Encompass_Preferred
YM-341619 is a potent, orally active small-molecule STAT6 inhibitor supplied for research use. It exhibits sub-nanomolar potency against STAT6 and is used in studies of allergic disease, Th2 differentiation, and JAK/STAT signaling. Supplied as a solid powder with manufacturer-recommended storage conditions for stability.
  • Potent STAT6 inhibition (IC50 0.70 nM).
  • High purity (98.01%).
  • Solid powder form for easy handling and formulation.
  • Stable under recommended storage conditions for long-term use.
  • Suitable for studies of allergic disease and Th2 differentiation.
  • Available in milligram-scale quantities for preclinical research.
Catalog No. 50-002-61283 Supplier Medchemexpress LLC Supplier No. HY13477110MG
YM-341619 is a potent, orally active small-molecule STAT6 inhibitor supplied for research use. It exhibits sub-nanomolar potency against STAT6 and is used in studies of allergic disease, Th2 differentiation, and JAK/STAT signaling. Supplied as a solid powder with manufacturer-recommended storage conditions for stability.
  • Potent STAT6 inhibition (IC50 0.70 nM).
  • High purity (98.01%).
  • Solid powder form for easy handling and formulation.
  • Stable under recommended storage conditions for long-term use.
  • Suitable for studies of allergic disease and Th2 differentiation.
  • Available in milligram-scale quantities for preclinical research.