Learn More
TARGETMOL CHEMICALS INC YM 534 5MG
SDP
Catalog No. 502576924
Encompass_Preferred
Also available in 1mg 10mg 25mg 50mg 100mg and bulk. Please contact Fisher for quotes. YM 534 has antitumor activity and platelet aggregation activity inhibits photoaffinity labeling of deglycosylated P-glycoprotein and can be used to study leukemia. purity: 99%
Catalog No. 50-257-6924 Supplier TARGETMOL CHEMICALS INC Supplier No. T352625MG
Also available in 1mg 10mg 25mg 50mg 100mg and bulk. Please contact Fisher for quotes. YM 534 has antitumor activity and platelet aggregation activity inhibits photoaffinity labeling of deglycosylated P-glycoprotein and can be used to study leukemia. purity: 99%