Learn More
Medchemexpress LLC Ym-53601 | 182959-33-7 | 99.4% | 372.86 g·mol⁻¹ | C21H22ClFN2O | 1 MG
SDP
Catalog No. 5000214385
Encompass_Preferred
YM-53601 is a small-molecule squalene synthase (FDFT1) inhibitor used in biochemical and pharmacology research to modulate cholesterol and triglyceride biosynthesis. It has demonstrated lipid-lowering effects in vivo and is supplied as a solid or as ready-to-use DMSO solutions for laboratory assays.
  • Inhibits squalene synthase (FDFT1) with reported IC50 ≈ 79 nM.
  • Reduces plasma cholesterol and triglyceride levels in animal studies.
  • Useful for studying lipid metabolism and hepatitis C virus propagation.
  • Available as solid and 10 mM DMSO solution for assay flexibility.
  • High purity suitable for biochemical and pharmacological assays.
  • Comes with QC documents such as data sheet, SDS, COA, HNMR, and LCMS.
Catalog No. 50-002-14385 Supplier Medchemexpress LLC Supplier No. HY100313A1MG
YM-53601 is a small-molecule squalene synthase (FDFT1) inhibitor used in biochemical and pharmacology research to modulate cholesterol and triglyceride biosynthesis. It has demonstrated lipid-lowering effects in vivo and is supplied as a solid or as ready-to-use DMSO solutions for laboratory assays.
  • Inhibits squalene synthase (FDFT1) with reported IC50 ≈ 79 nM.
  • Reduces plasma cholesterol and triglyceride levels in animal studies.
  • Useful for studying lipid metabolism and hepatitis C virus propagation.
  • Available as solid and 10 mM DMSO solution for assay flexibility.
  • High purity suitable for biochemical and pharmacological assays.
  • Comes with QC documents such as data sheet, SDS, COA, HNMR, and LCMS.