Learn More
Cayman Chemical YM-53601, 182959-33-7, 500 µg
SDP
Supplier:  Cayman Chemical 18113500
Encompass_Preferred
Inhibits squalene synthase activity in rat hepatic microsomes and human Hep-G2 cells (IC50 values of 90 and 79 nM, respectively); inhibits cholesterol biosynthesis in rats reducing both cholesterol and triglyceride levels in plasma
Catalog No. NC0701380
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.