Learn More
Apexbio Technology LLC YM 58483,10mg CAS# 223499-30-7
SDP
Catalog No. 502240212
Encompass_Preferred
YM 58483, also known as BTP2, is a SOCE blocker. SOCE has been found in macrophages, basophils, dendritic cells, and mast cells as well as T cells. The SOCE-related channels include the Ca2+ release-activated Ca2+ (CRAC) channel which is a highly Ca2+-selective channel, and the non-selective, Ca2+-permeable transient receptor potential (TRP) channels, which regulate the activation of non-excitable cells such as lymphocytes. YM 58483 has been reported to inhibit cytokine production and proliferation in T cells, and may serve as a potential medicinal candidate for treatment of bronchial asthma.  Other sizes are also available. Please inqury us for quote.
This item is not returnable. View return policy
Catalog No. 50-224-0212 Supplier Apexbio Technology LLC Supplier No. B754210
YM 58483, also known as BTP2, is a SOCE blocker. SOCE has been found in macrophages, basophils, dendritic cells, and mast cells as well as T cells. The SOCE-related channels include the Ca2+ release-activated Ca2+ (CRAC) channel which is a highly Ca2+-selective channel, and the non-selective, Ca2+-permeable transient receptor potential (TRP) channels, which regulate the activation of non-excitable cells such as lymphocytes. YM 58483 has been reported to inhibit cytokine production and proliferation in T cells, and may serve as a potential medicinal candidate for treatment of bronchial asthma.  Other sizes are also available. Please inqury us for quote.