Learn More
Medchemexpress LLC HY-100831 5mg , YM-58483 CAS:223499-30-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1008315MG
Encompass_Preferred
HY-100831 5mg YM-58483 CAS: 223499-30-7 YM-58483 (BTP2) is the first selective and potent inhibitor of CRAC channels and subsequent Ca2+ signals. YM-584832 is a blocker of store-operated Ca2+ entry (SOCE). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0385