Learn More
Selleck Chemical LLC YM-58483 (BTP2) 100mg 223499-30-7
SDP
Supplier:  Selleck Chemical LLC S8380100MG
Encompass_Preferred
YM-58483/BTP2 is a blocker of store-operated Ca2+ entry (SOCE), which regulates the activation of non-excitable cells such as lymphocytes. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-6013