Learn More
Selleck Chemical LLC YM 90709
SDP
Catalog No. 5000002859
Encompass_Preferred
YM 90709 is a novel IL-5 inhibitor which selectively blocks the binding of IL-5 to the IL-5 receptor (IL-5R)
Catalog No. 50-000-02859 Supplier Selleck Chemical LLC Supplier No. S57645MG
YM 90709 is a novel IL-5 inhibitor which selectively blocks the binding of IL-5 to the IL-5 receptor (IL-5R)