Learn More
Teknova YM Agar Plates. 100mm Plates, 20 Plates per Sleeve, Sterile. 4 pack.
SDP
Catalog No. 502387354
Encompass
0.3% Malt Extract, 0.3% Yeast Extract, 0.5% Peptone, 1.0% Dextrose, 2.0% Agar
Catalog No. 50-238-7354 Supplier Teknova Supplier No. Y20104PK
0.3% Malt Extract, 0.3% Yeast Extract, 0.5% Peptone, 1.0% Dextrose, 2.0% Agar