Learn More
Teknova YM Broth. 1000mL, Sterile 2 pack.
SDP
Catalog No. 502387271
Encompass
0.3% Yeast Extract, 0.5% Tryptone,0.3% Malt Extract, 1.0% Cerelose Anhydrous Dextrose (Glucose)
Catalog No. 50-238-7271 Supplier Teknova Supplier No. Y07312PK
0.3% Yeast Extract, 0.5% Tryptone,0.3% Malt Extract, 1.0% Cerelose Anhydrous Dextrose (Glucose)