Learn More
Bioworld YM Broth, 500 g
Catalog No. 50254488
Encompass_Preferred
YM BrothUse for the cultivation of yeasts, molds and other aciduric microorganismspH: 6.2Dehydrated Appearance: beige, homogeneous powderPrepared Appearance: yellow to gold brothUse 21g per literSoluble in waterContains (g/L):Bacto peptone-5gYeast extract-3gMalt extract-3gDextrose -10g
Catalog No. 50-254-488 Supplier Bioworld Supplier No. 306260161
YM BrothUse for the cultivation of yeasts, molds and other aciduric microorganismspH: 6.2Dehydrated Appearance: beige, homogeneous powderPrepared Appearance: yellow to gold brothUse 21g per literSoluble in waterContains (g/L):Bacto peptone-5gYeast extract-3gMalt extract-3gDextrose -10g