Learn More
Medchemexpress LLC HY-103355 5mg Medchemexpress, YM022 CAS:145084-28-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1033555MG
Encompass_Preferred
Medchemexpress, HY-103355 5mg YM022 CAS:145084-28-2 YM022 is a highly potent, selective and orally active gastrin/cholecystokinin (CCK)-B receptor (CCK-BR) antagonist. YM022 shows the Ki values of 68 pM and 63 nM for CCK-B and CCK-A receptor, respectively. YM022 can inhibit gastrin-induced gastric acid secretion and histidine decarboxylase activation in vivo[3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8084