Learn More
Selleck Chemical LLC YM155 (Sepantronium Bromide) 100mg 781661-94-7
SDP
Catalog No. 508911
Encompass_Preferred
YM155 (Sepantronium Bromide) is a potent survivin suppressant by inhibiting Survivin promoter activity with IC50 of 0.54 nM in HeLa-SURP-luc and CHO-SV40-luc cells; does not significantly inhibit SV40 promoter activity, but is observed to slightly inhibit the interaction of Survivin with XIAP. YM155 down-regulates survivin and XIAP, modulates autophagy and induces autophagy-dependent DNA damage in breast cancer cells. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-891-1 Supplier Selleck Chemical LLC Supplier No. S1130100MG
YM155 (Sepantronium Bromide) is a potent survivin suppressant by inhibiting Survivin promoter activity with IC50 of 0.54 nM in HeLa-SURP-luc and CHO-SV40-luc cells; does not significantly inhibit SV40 promoter activity, but is observed to slightly inhibit the interaction of Survivin with XIAP. YM155 down-regulates survivin and XIAP, modulates autophagy and induces autophagy-dependent DNA damage in breast cancer cells. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.