Learn More
Medchemexpress LLC HY-101627 1mg Medchemexpress, YM17E CAS:124900-72-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1016271MG
Encompass_Preferred
Medchemexpress, HY-101627 1mg YM17E CAS:124900-72-7 YM17E is an inhibitor of acyl CoA:cholesterol acyltransferase (ACAT), with IC50 of 44 nM in rabbit liver microsomes in vitro. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7863