Learn More
Abcam YM298198 hydrochloride, mGlu1 antagonist, 10MG
Catalog No. 502707940
Encompass_Preferred
MW 378.9 Da, Purity >99%. Potent, selective, water-soluble, non-competitive mGlu1 antagonist (IC50 = 16 nM) Selective over mGlu2,3,4,5,6, and 7 and ionotropic receptors (IC50 > 10 μM). Orally active analgesic *in vivo*. Also available in simple stock solutions (ab146667) - add 1 ml of water to get an exact, ready-to-use concentration.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7940 Supplier Abcam Supplier No. AB12001510MG
Add to Cart
Add to Cart
MW 378.9 Da, Purity >99%. Potent, selective, water-soluble, non-competitive mGlu1 antagonist (IC50 = 16 nM) Selective over mGlu2,3,4,5,6, and 7 and ionotropic receptors (IC50 > 10 μM). Orally active analgesic *in vivo*. Also available in simple stock solutions (ab146667) - add 1 ml of water to get an exact, ready-to-use concentration.
The product is subject to the following: Abcam Restricted Use Statement