Learn More
Cayman Chemical YM90KhydroCLOrID hydrate 10mg
SDP
Catalog No. 502989424
Encompass_Preferred
An AMPA receptor antagonist (Ki = 0.084 μM in a radioligand binding assay using rat brain membranes); selective for AMPA over kainate, NMDA-sensitive glutamate, and strychnine-insensitive glycine receptors (Kis = 2.2, >100, and 37 μM, respectively); intrastriatal administration inhibits AMPA-induced decreases in ChAT activity in rat striatum ex vivo; inhibits audiogenic seizures in susceptible DBA/2 mice with a minimum effective dose of 3 mg/kg, i.p.; intravenous infusion reduces infarct size in a rat model of focal cerebral ischemia induced by thrombotic MCAO in rats at 20 mg/kg per hour for 4 hours
Catalog No. 50-298-9424 Supplier Cayman Chemical Supplier No. 3068310 MG
An AMPA receptor antagonist (Ki = 0.084 μM in a radioligand binding assay using rat brain membranes); selective for AMPA over kainate, NMDA-sensitive glutamate, and strychnine-insensitive glycine receptors (Kis = 2.2, >100, and 37 μM, respectively); intrastriatal administration inhibits AMPA-induced decreases in ChAT activity in rat striatum ex vivo; inhibits audiogenic seizures in susceptible DBA/2 mice with a minimum effective dose of 3 mg/kg, i.p.; intravenous infusion reduces infarct size in a rat model of focal cerebral ischemia induced by thrombotic MCAO in rats at 20 mg/kg per hour for 4 hours