Learn More
YMC America, Inc YMCbasic 3um 5x0.5mm 1/32
SDP
Catalog No. 5000879521
Encompass
YMCbasic is a unique reversed-phase column designed for analysis of pharmaceuticals and basic compounds Innovative silica synthesis and state-of-the-art silane bonding result in a chromatographic material with neutral silanols This minimizes interactions with both basic and acidic compounds
Catalog No. 50-008-79521 Supplier YMC America, Inc Supplier No. BA99S03E5J0RU
YMCbasic is a unique reversed-phase column designed for analysis of pharmaceuticals and basic compounds Innovative silica synthesis and state-of-the-art silane bonding result in a chromatographic material with neutral silanols This minimizes interactions with both basic and acidic compounds