Learn More
YMC America, Inc YMCbasic 5um 75x0.3mm 1/32
SDP
Catalog No. 5000879527
Encompass
YMCbasic is a unique reversed-phase column designed for analysis of pharmaceuticals and basic compounds Innovative silica synthesis and state-of-the-art silane bonding result in a chromatographic material with neutral silanols This minimizes interactions with both basic and acidic compounds
Catalog No. 50-008-79527 Supplier YMC America, Inc Supplier No. BA99S05L5H0RU
Add to Cart
Add to Cart
YMCbasic is a unique reversed-phase column designed for analysis of pharmaceuticals and basic compounds Innovative silica synthesis and state-of-the-art silane bonding result in a chromatographic material with neutral silanols This minimizes interactions with both basic and acidic compounds