Learn More
YMC America, Inc YMCbasic col 10um 150x10mm
SDP
Catalog No. 5000877110
Encompass_Preferred
YMCbasic is a unique reversed-phase column designed for analysis of pharmaceuticals and basic compounds Innovative silica synthesis and state-of-the-art silane bonding result in a chromatographic material with neutral silanols This minimizes interactions with both basic and acidic compounds
Catalog No. 50-008-77110 Supplier YMC America, Inc Supplier No. BA99S111510WT
YMCbasic is a unique reversed-phase column designed for analysis of pharmaceuticals and basic compounds Innovative silica synthesis and state-of-the-art silane bonding result in a chromatographic material with neutral silanols This minimizes interactions with both basic and acidic compounds