Learn More
YMC America, Inc YMCbasic col 10um 250x4.6mm
SDP
Catalog No. 5000876903
Encompass_Preferred
YMCbasic is a unique reversed-phase column designed for analysis of pharmaceuticals and basic compounds Innovative silica synthesis and state-of-the-art silane bonding result in a chromatographic material with neutral silanols This minimizes interactions with both basic and acidic compounds
This item is not returnable. View return policy
Catalog No. 50-008-76903 Supplier YMC America, Inc Supplier No. BA99S112546WT
YMCbasic is a unique reversed-phase column designed for analysis of pharmaceuticals and basic compounds Innovative silica synthesis and state-of-the-art silane bonding result in a chromatographic material with neutral silanols This minimizes interactions with both basic and acidic compounds