Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ YNB with Ammonium Sulfate (Powder)

Catalog No. MP114027512 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
227 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP114027512 227 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP114027512 Supplier MP Biomedicals™ Supplier No. 114027512
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Causes vigorous growth of Saccharomyces cerevisiae. MP Biomedicals YNB with Ammonium Sulfate (Powder) is a well-defined composition of salts, vitamins and a nitrogen source. It is a component of synthetic defined yeast media.

TRUSTED_SUSTAINABILITY

Specifications

Form Powder
For Use With (Application) YNB w/ Ammonium Sulfate is a component of synthetic defined yeast media.
Quantity 227 g
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.