Learn More
Medchemexpress LLC Ynt-185 | 1804978-81-1 | 98.5% | 615.74 | 1 ML
SDP
Catalog No. 5000349990
Encompass_Preferred
YNT-185 is a nonpeptide, selective orexin type-2 receptor (OX2R) agonist. It has EC50s of 0.028 μM for OX2R and 2.75 μM for OX1R. YNT-185 has been shown to ameliorate narcolepsy-cataplexy symptoms in mouse models. It is for research use only and not sold to patients.
  • Selectively activates the orexin type-2 receptor
  • Ameliorates narcolepsy-cataplexy symptoms in mouse models
  • Increases wakefulness in mice
  • Significantly increases wake time in a dose-dependent manner
Catalog No. 50-003-49990 Supplier Medchemexpress LLC Supplier No. HY13618110MM/1ML
YNT-185 is a nonpeptide, selective orexin type-2 receptor (OX2R) agonist. It has EC50s of 0.028 μM for OX2R and 2.75 μM for OX1R. YNT-185 has been shown to ameliorate narcolepsy-cataplexy symptoms in mouse models. It is for research use only and not sold to patients.
  • Selectively activates the orexin type-2 receptor
  • Ameliorates narcolepsy-cataplexy symptoms in mouse models
  • Increases wakefulness in mice
  • Significantly increases wake time in a dose-dependent manner