Learn More
TARGETMOL CHEMICALS INC YNT-185 5MG
SDP
Catalog No. 502580065
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. YNT-185 is a nonpeptide selective agonist of the orexin type-2 receptor (OX2R) demonstrating EC50 values of 0.028 uM and 2.75 uM for OX2R and OX1R respectively. Furthermore YNT-185 effectively alleviates symptoms of narcolepsy-cataplexy in mouse models. purity: 98%
Catalog No. 50-258-0065 Supplier TARGETMOL CHEMICALS INC Supplier No. T392295MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. YNT-185 is a nonpeptide selective agonist of the orexin type-2 receptor (OX2R) demonstrating EC50 values of 0.028 uM and 2.75 uM for OX2R and OX1R respectively. Furthermore YNT-185 effectively alleviates symptoms of narcolepsy-cataplexy in mouse models. purity: 98%