Learn More
Medchemexpress LLC YNT-185 dihydrochloride | 1804978-82-2 | 1804978-82-2 | 99.8% | 688.66 | 100 MG
SDP
Catalog No. 5000348737
Encompass_Preferred
YNT-185 dihydrochloride is a nonpeptide, selective orexin type-2 receptor (OX2R) agonist, with EC50 values of 0.028 μM for OX2R and 2.75 μM for OX1R. It ameliorates narcolepsy-cataplexy symptoms in mouse models and increases wakefulness in mice.
  • Selective OX2R agonist
  • Ameliorates narcolepsy-cataplexy symptoms
  • Increases wakefulness
Catalog No. 50-003-48737 Supplier Medchemexpress LLC Supplier No. HY136181100MG
YNT-185 dihydrochloride is a nonpeptide, selective orexin type-2 receptor (OX2R) agonist, with EC50 values of 0.028 μM for OX2R and 2.75 μM for OX1R. It ameliorates narcolepsy-cataplexy symptoms in mouse models and increases wakefulness in mice.
  • Selective OX2R agonist
  • Ameliorates narcolepsy-cataplexy symptoms
  • Increases wakefulness