Learn More
Medchemexpress LLC YNT-185 dihydrochloride | 1804978-82-2 | 99.8% | 688.66 | 25 MG
SDP
Catalog No. 5000352305
Encompass_Preferred
YNT-185 dihydrochloride is a nonpeptide, selective orexin type-2 receptor (OX2R) agonist. It demonstrates high selectivity with EC50s of 0.028 μM for OX2R and 2.75 μM for OX1R. This compound has been shown to ameliorate narcolepsy-cataplexy symptoms in mouse models, increasing wakefulness and decreasing NREM sleep time.
  • Selective orexin type-2 receptor (OX2R) agonist
  • Nonpeptide compound
  • EC50 of 0.028 μM for OX2R
  • Ameliorates narcolepsy-cataplexy symptoms in mouse models
  • Increases wakefulness
  • Decreases NREM sleep time
Catalog No. 50-003-52305 Supplier Medchemexpress LLC Supplier No. 25MGHY136181
YNT-185 dihydrochloride is a nonpeptide, selective orexin type-2 receptor (OX2R) agonist. It demonstrates high selectivity with EC50s of 0.028 μM for OX2R and 2.75 μM for OX1R. This compound has been shown to ameliorate narcolepsy-cataplexy symptoms in mouse models, increasing wakefulness and decreasing NREM sleep time.
  • Selective orexin type-2 receptor (OX2R) agonist
  • Nonpeptide compound
  • EC50 of 0.028 μM for OX2R
  • Ameliorates narcolepsy-cataplexy symptoms in mouse models
  • Increases wakefulness
  • Decreases NREM sleep time