Learn More
Medchemexpress LLC Ynt-185 dihydrochloride | 1804978-82-2 | 99.8% | C33H39Cl2N5O5S | 10MG
SDP
Catalog No. 5000261789
Encompass_Preferred
YNT-185 dihydrochloride is a nonpeptide, selective orexin type-2 receptor (OX2R) agonist used as a research tool to probe orexin signaling and sleep disorder models. It exhibits high potency at OX2R (EC50 = 0.028 μM) and lower activity at OX1R, and is supplied as a solid or as a 10 mM DMSO stock solution for laboratory use. For research use only.
  • Selective OX2R agonist with nanomolar potency (EC50 = 0.028 μM).
  • Lower activity at OX1R, enabling receptor subtype studies.
  • High purity for reproducible biochemical and pharmacological results.
  • Available in multiple pack sizes and as a 10 mM DMSO stock solution.
  • Supplied as an off-white to light-yellow solid suitable for formulation.
Catalog No. 50-002-61789 Supplier Medchemexpress LLC Supplier No. HY13618110MG
YNT-185 dihydrochloride is a nonpeptide, selective orexin type-2 receptor (OX2R) agonist used as a research tool to probe orexin signaling and sleep disorder models. It exhibits high potency at OX2R (EC50 = 0.028 μM) and lower activity at OX1R, and is supplied as a solid or as a 10 mM DMSO stock solution for laboratory use. For research use only.
  • Selective OX2R agonist with nanomolar potency (EC50 = 0.028 μM).
  • Lower activity at OX1R, enabling receptor subtype studies.
  • High purity for reproducible biochemical and pharmacological results.
  • Available in multiple pack sizes and as a 10 mM DMSO stock solution.
  • Supplied as an off-white to light-yellow solid suitable for formulation.