Learn More
Medchemexpress LLC HY-13526 25mg , YO-01027 Dibenzazepine;DBZ CAS:209984-56-5 Purity:98%
SDP
Catalog No. 501493990
Encompass_Preferred
Medchemexpress, HY-13526 25mg YO-01027 Dibenzazepine;DBZ CAS:209984-56-5 Formula:C26H23F2N3O3 IC50: 2.92±0.22 (Notch), 2.64±0.30 (APPL) nM Purity:98% YO-01027 (Dibenzazepine;DBZ) is a potent γ-secretase inhibitor with IC 50 values of 2.92±0.22 and 2.64±0.30 nM for Notch and APPL cleavage, respectively. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-3990 Supplier Medchemexpress LLC Supplier No. HY1352625MG
Medchemexpress, HY-13526 25mg YO-01027 Dibenzazepine;DBZ CAS:209984-56-5 Formula:C26H23F2N3O3 IC50: 2.92±0.22 (Notch), 2.64±0.30 (APPL) nM Purity:98% YO-01027 (Dibenzazepine;DBZ) is a potent γ-secretase inhibitor with IC 50 values of 2.92±0.22 and 2.64±0.30 nM for Notch and APPL cleavage, respectively. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.