Learn More
Apexbio Technology LLC YO-01027 (Dibenzazepine, DBZ) 209984-56-5 10mM (in 1mL DMSO)
SDP
Catalog No. 5000063965
Encompass_Preferred
YO-01027 (Dibenzazepine DBZ CAS 209984-56-5) is a potent inhibitor targeting -secretase a multimeric aspartyl protease involved in the proteolytic cleavage of type I integral membrane proteins notably amyloid precursor protein-like (APPL) proteins and Notch receptors YO-01027 interacts with the N-terminal regions of presenilin dose-dependently inhibiting cleavage of APPL and Notch proteins Research indicates that YO-01027 modulates cell differentiation by suppressing Notch signaling pathways influencing cellular transitions and mucin (MUC16) synthesis illustrating its utility in cell biology and therapeutic studies of Notch-related signaling processes
Catalog No. 50-000-63965 Supplier Apexbio Technology LLC Supplier No. A40185.1
YO-01027 (Dibenzazepine DBZ CAS 209984-56-5) is a potent inhibitor targeting -secretase a multimeric aspartyl protease involved in the proteolytic cleavage of type I integral membrane proteins notably amyloid precursor protein-like (APPL) proteins and Notch receptors YO-01027 interacts with the N-terminal regions of presenilin dose-dependently inhibiting cleavage of APPL and Notch proteins Research indicates that YO-01027 modulates cell differentiation by suppressing Notch signaling pathways influencing cellular transitions and mucin (MUC16) synthesis illustrating its utility in cell biology and therapeutic studies of Notch-related signaling processes