Learn More
Medchemexpress LLC Yoda 1 | 448947-81-7 | MFCD09839937 | ≥98% | 421.32 | 250 MG
SDP
Catalog No. 5000384434
Encompass_Preferred
Yoda 1 is a potent and selective Piezo1 agonist that activates purified Piezo1 channels. It potently inhibits macropinocytosis induced by epidermal growth factor (EGF) and enhances Ca2+ influx. This is followed by the activation of the calcium-activated potassium channel KCa3.1 and inhibition of Rac1 activation.
  • Activates purified Piezo1 channels
  • Potently inhibits macropinocytosis induced by epidermal growth factor (EGF)
  • Enhances Ca2+ influx
  • Activates the calcium-activated potassium channel KCa3.1
  • Inhibits Rac1 activation
Catalog No. 50-003-84434 Supplier Medchemexpress LLC Supplier No. 250MGHY18723
Yoda 1 is a potent and selective Piezo1 agonist that activates purified Piezo1 channels. It potently inhibits macropinocytosis induced by epidermal growth factor (EGF) and enhances Ca2+ influx. This is followed by the activation of the calcium-activated potassium channel KCa3.1 and inhibition of Rac1 activation.
  • Activates purified Piezo1 channels
  • Potently inhibits macropinocytosis induced by epidermal growth factor (EGF)
  • Enhances Ca2+ influx
  • Activates the calcium-activated potassium channel KCa3.1
  • Inhibits Rac1 activation