Learn More
Selleck Chemical LLC Yoda1 5mg
SDP
Supplier:  Selleck Chemical LLC S66785MG
Encompass_Preferred
Yoda1 acts as an agonist for both human and mouse Piezo1(EC50 of 17.1 and 26.6 ?M for mouse and human). Yoda1 activates purified Piezo1 channels in the absence of other cellular components. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3206